accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3906-16-9

IUPAC Name: 1-(naphthalen-2-yl)ethan-1-amine
Synonyms: 1-(naphthalen-2-yl)ethanamine
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: KHSYYLCXQKCYQX-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results