accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39065-54-8

IUPAC Name: 6-phenylpyridine-3-carbonitrile
Synonyms: 6-phenylpyridine-3-carbonitrile
Molecular Weight (g/mol): 180.21
Molecular Formula: C12H8N2
InChi Key: OSRYUVXCTNUGRM-UHFFFAOYSA-N
SMILES: N#CC1=CN=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results