CAS RN 391-53-7
CAS RN 391-53-7
IUPAC Name:
2-fluoro-9H-carbazole
Synonyms:
2-fluoro-9H-carbazole
Molecular Weight (g/mol):
185.2
Molecular Formula:
C12H8FN
InChi Key:
RQGSXFCTBTUNRR-UHFFFAOYSA-N
SMILES:
FC1=CC2=C(C=C1)C1=C(N2)C=CC=C1
1
–
1
of
1
results
1
2-Fluoro-9H-carbazole 98.0+%, TCI America™

CAS: 391-53-7 Molecular Formula: C12H8FN Molecular Weight (g/mol): 185.20 MDL Number: MFCD09033508 InChI Key: RQGSXFCTBTUNRR-UHFFFAOYSA-N PubChem CID: 11816279 IUPAC Name: 2-fluoro-9H-carbazole SMILES: FC1=CC2=C(C=C1)C1=C(N2)C=CC=C1