accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3913-02-8

IUPAC Name: 2-butyloctan-1-ol
Synonyms: butyloctanol
Molecular Weight (g/mol): 186.34
Molecular Formula: C12H26O
InChi Key: XMVBHZBLHNOQON-UHFFFAOYNA-N
SMILES: CCCCCCC(CO)CCCC
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results