accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39145-52-3

IUPAC Name: 2-acetamido-3-(1H-imidazol-5-yl)propanoic acid hydrate
Synonyms: 2-acetamido-3-(3H-imidazol-4-yl)propanoic acid hydrate
Molecular Weight (g/mol): 215.21
Molecular Formula: C8H13N3O4
InChi Key: PSWSDQRXCOJSFC-UHFFFAOYNA-N
SMILES: O.CC(=O)NC(CC1=CN=CN1)C(O)=O
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)