accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39151-19-4

IUPAC Name: 1-(3,5-dimethoxyphenyl)ethan-1-one
Synonyms: 1-(3,5-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: YJKHOUIVWKQRSL-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results