CAS RN 39160-31-1
CAS RN 39160-31-1
IUPAC Name:
4-fluoropyridine hydrochloride
Synonyms:
4-fluoropyridine hydrochloride
Molecular Weight (g/mol):
133.55
Molecular Formula:
C5H5ClFN
InChi Key:
QRUIPALQRYLTAF-UHFFFAOYSA-N
SMILES:
Cl.FC1=CC=NC=C1
1
–
1
of
1
results
1
4-Fluoropyridine Hydrochloride 98.0+%, TCI America™

CAS: 39160-31-1 Molecular Formula: C5H5ClFN Molecular Weight (g/mol): 133.55 MDL Number: MFCD00234055 InChI Key: QRUIPALQRYLTAF-UHFFFAOYSA-N PubChem CID: 24820326 IUPAC Name: 4-fluoropyridine;hydrochloride SMILES: C1=CN=CC=C1F.Cl