CAS RN 3919-74-2
IUPAC Name:
3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Synonyms:
3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol):
255.63
Molecular Formula:
C11H7ClFNO3
InChi Key:
REIPZLFZMOQHJT-UHFFFAOYSA-N
SMILES:
CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl
1
–
1
of
1
results
1
Promotions Available
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 99%