accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3919-76-4

IUPAC Name: 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Synonyms: 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 272.08
Molecular Formula: C11H7Cl2NO3
InChi Key: WQXUUMUOERZZAE-UHFFFAOYSA-N
SMILES: CC1=C(C(O)=O)C(=NO1)C1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results