accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39190-67-5

IUPAC Name: (butan-2-yl)(propyl)amine
Synonyms: propyl sec-butylamine
Molecular Weight (g/mol): 115.22
Molecular Formula: C7H17N
InChi Key: QYNZYUUXSVZDJO-UHFFFAOYNA-N
SMILES: CCCNC(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results