accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOONSH3CH3CH3CCH3(E)

CAS RN 39196-18-4

IUPAC Name: (E)-[3,3-dimethyl-1-(methylsulfanyl)butan-2-ylidene]amino N-methylcarbamate
Synonyms: thiofanox
Molecular Weight (g/mol): 218.32
Molecular Formula: C9H18N2O2S
InChi Key: FZSVSABTBYGOQH-XFFZJAGNSA-N
SMILES: CNC(=O)O\N=C(\CSC)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results