accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 392-13-2

H3CHNF

CAS RN 392-13-2

IUPAC Name: 5-fluoro-3-methyl-1H-indole
Synonyms: 5-fluoro-3-methyl-1H-indole
Molecular Weight (g/mol): 149.17
Molecular Formula: C9H8FN
InChi Key: CSDHAGJNOQIBHZ-UHFFFAOYSA-N
SMILES: CC1=CNC2=CC=C(F)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)