accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFF

CAS RN 392-56-3

IUPAC Name: hexafluorobenzene
Synonyms: hexafluorobenzene
Molecular Weight (g/mol): 186.06
Molecular Formula: C6F6
InChi Key: ZQBFAOFFOQMSGJ-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (5)
  • (7)
  • (3)
Boiling Point 
  • (3)
  • (5)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (1)
  • (3)
  • (6)
  • (5)
  • (7)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (7)
  • (8)
  • (1)