CAS RN 392-63-2
IUPAC Name:
N,N-diethyl-2,2,3,3-tetrafluoropropanamide
Synonyms:
N,N-diethyl-2,2,3,3-tetrafluoropropanamide
Molecular Weight (g/mol):
201.17
Molecular Formula:
C7H11F4NO
InChi Key:
OIZFBXRWENZGCS-UHFFFAOYSA-N
SMILES:
CCN(CC)C(=O)C(F)(F)C(F)F