accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNO

CAS RN 3920-50-1

IUPAC Name: 1H-pyrazole-5-carbaldehyde
Synonyms: 2H-pyrazole-3-carbaldehyde
Molecular Weight (g/mol): 96.09
Molecular Formula: C4H4N2O
InChi Key: ICFGFAUMBISMLR-UHFFFAOYSA-N
SMILES: O=CC1=CC=NN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results