accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39255-32-8

IUPAC Name: ethyl 2-methylpentanoate
Synonyms: manzanate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: HZPKNSYIDSNZKW-UHFFFAOYNA-N
SMILES: CCCC(C)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results