accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39255-60-2

IUPAC Name: N'-(4-bromo-2-cyanophenyl)-N,N-dimethylmethanimidamide
Synonyms: N'-(4-bromo-2-cyanophenyl)-N,N-dimethylmethanimidamide
Molecular Weight (g/mol): 252.12
Molecular Formula: C10H10BrN3
InChi Key: ZGEYANFICVYJSY-UHFFFAOYSA-N
SMILES: CN(C)C=NC1=C(C=C(Br)C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results