accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BrN

CAS RN 39263-32-6

IUPAC Name: 2-amino-5-bromobenzonitrile
Synonyms: 2-amino-5-bromobenzonitrile
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: OATYCBHROMXWJO-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1C#N
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)