accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOO

CAS RN 392659-97-1

IUPAC Name: 5-[(morpholin-4-yl)methyl]furan-2-carbaldehyde
Synonyms: 5-(morpholin-4-ylmethyl)furan-2-carbaldehyde
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: ZQILSGYYJOBENS-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2CCOCC2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results