accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 392660-27-4

IUPAC Name: 1-(2-fluoro-4-nitrophenyl)-1,4-diazepane
Synonyms: 1-(2-fluoro-4-nitrophenyl)-1,4-diazepane
Molecular Weight (g/mol): 239.25
Molecular Formula: C11H14FN3O2
InChi Key: BSSBZGVNKLRQEL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results