accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39269-10-8

IUPAC Name: adamantane-1,3-dicarboxylate
Synonyms: adamantane-1,3-dicarboxylate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: PAVQGHWQOQZQEH-UHFFFAOYSA-L
SMILES: [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)