accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39270-39-8

IUPAC Name: (2,3-dihydro-1,4-benzodioxin-6-yl)methanol
Synonyms: 2,3-dihydro-1,4-benzodioxin-6-ylmethanol
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: FFLHNBGNAWYMRH-UHFFFAOYSA-N
SMILES: OCC1=CC=C2OCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results