accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaBrPtBrBrBrBrBr

CAS RN 39277-13-9

IUPAC Name: disodium hexabromoplatinumdiuide
Synonyms: disodium hexabromoplatinumdiuide
Molecular Weight (g/mol): 720.49
Molecular Formula: Br6Na2Pt
InChi Key: RFZKZIKSQSDBBI-UHFFFAOYSA-H
SMILES: [Na+].[Na+].Br[Pt--](Br)(Br)(Br)(Br)Br
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results