CAS RN 3930-83-4
IUPAC Name:
2-iodobenzamide
Synonyms:
2-iodobenzamide
Molecular Weight (g/mol):
247.04
Molecular Formula:
C7H6INO
InChi Key:
YEOYYWCXWUDVCX-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=CC=C1I
1
–
1
of
1
results
1
2-Iodobenzamide 98.0+%, TCI America™
CAS: 3930-83-4 Molecular Formula: C7H6INO Molecular Weight (g/mol): 247.035 MDL Number: MFCD00099248 InChI Key: YEOYYWCXWUDVCX-UHFFFAOYSA-N PubChem CID: 77529 IUPAC Name: 2-iodobenzamide SMILES: C1=CC=C(C(=C1)C(=O)N)I