accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFHO

CAS RN 394-32-1

IUPAC Name: 1-(5-fluoro-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(5-fluoro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: KOFFXZYMDLWRHX-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=CC=C1O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)