accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOF

CAS RN 394-35-4

IUPAC Name: methyl 2-fluorobenzoate
Synonyms: methyl 2-fluorobenzoate
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: QAFJIJWLEBLXHH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results