accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 394-46-7

IUPAC Name: 1-ethenyl-2-fluorobenzene
Synonyms: 1-ethenyl-2-fluorobenzene
Molecular Weight (g/mol): 122.14
Molecular Formula: C8H7F
InChi Key: YNQXOOPPJWSXMW-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C=C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)