accessibility menu, dialog, popup

UserName

Viewing profile as user:
FHOO

CAS RN 394-50-3

IUPAC Name: 3-fluoro-2-hydroxybenzaldehyde
Synonyms: 3-fluoro-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: NWDHTEIVMDYWQJ-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=CC=C1F
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)