accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFFF

CAS RN 394-59-2

IUPAC Name: 2,2,2-trifluoro-1-(4-methylphenyl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(4-methylphenyl)ethanone
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: DYILUJUELMWXAL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)