accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HOH3COOHOHOOHO

CAS RN 39413-05-3

IUPAC Name: 2-hydroxy-2-(2-oxopropyl)butanedioic acid; propan-2-ol
Synonyms: 2-hydroxy-2-(2-oxopropyl)butanedioic acid; isopropyl alcohol
Molecular Weight (g/mol): 250.25
Molecular Formula: C10H18O7
InChi Key: XBLJCYOUYPSETL-UHFFFAOYNA-N
SMILES: CC(C)O.CC(=O)CC(O)(CC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results