CAS RN 39430-27-8
IUPAC Name:
nickel(2+) tetrahydrate hydroxide carbonate
Synonyms:
nickel(2+) tetrahydrate hydroxide carbonate
Molecular Weight (g/mol):
207.77
Molecular Formula:
CH9NiO8
InChi Key:
KNXPLWTUEQLSEP-UHFFFAOYSA-K
SMILES:
O.O.O.O.[OH-].[Ni++].[O-]C([O-])=O
1
–
2
of
2
results
1
2
Promotions Available
Nickel Carbonate, Basic, Tetrahydrate, Powder, 45%, Spectrum™ Chemical
Nickel Carbonate, Basic, Tetrahydrate, Powder, 45%, Spectrum™ Chemical
CAS: 39430-27-8