accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHNHO(Z)(Z)

CAS RN 3946-91-6

IUPAC Name: (6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)phenyl]amino}methylidene)cyclohexa-2,4-dien-1-one
Synonyms: (6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)phenyl]amino}methylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 316.36
Molecular Formula: C20H16N2O2
InChi Key: MGJWLVOAKUZEGP-VMNXYWKNSA-N
SMILES: O=C1C=CC=C\C1=C\NC1=CC=CC=C1N\C=C1\C=CC=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results