accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39478-78-9

CH3BrH2N

CAS RN 39478-78-9

IUPAC Name: 5-bromo-2-methylaniline
Synonyms: 3-bromo-6-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: RXQNKKRGJJRMKD-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results