accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39493-62-4

IUPAC Name: methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate
Synonyms: methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate
Molecular Weight (g/mol): 184.19
Molecular Formula: C9H12O4
InChi Key: SEBIQMPNDQLYOX-UHFFFAOYNA-N
SMILES: COC(=O)C1C(C)CC(=O)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results