accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOFFF

CAS RN 395-64-2

IUPAC Name: 2,5-bis(trifluoromethyl)benzaldehyde
Synonyms: 2,5-bis(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 242.12
Molecular Formula: C9H4F6O
InChi Key: PICGJIQKPLBIES-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C(C=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results