accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 395-81-3

IUPAC Name: 5-fluoro-2-nitrobenzaldehyde
Synonyms: 5-fluoro-2-nitrobenzaldehyde
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H4FNO3
InChi Key: KKAFVHUJZPVWND-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C=C1C=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)