CAS RN 3951-10-8
IUPAC Name:
2-(4-methoxyphenyl)acetyl 2-(4-methoxyphenyl)acetate
Synonyms:
2-(4-methoxyphenyl)acetyl 2-(4-methoxyphenyl)acetate
Molecular Weight (g/mol):
314.34
Molecular Formula:
C18H18O5
InChi Key:
KYCAEIXHUXBNTQ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CC(=O)OC(=O)CC2=CC=C(OC)C=C2)C=C1
1
–
1
of
1
results
1
4-Methoxyphenylacetic Anhydride 98.0+%, TCI America™
CAS: 3951-10-8 Molecular Formula: C18H18O5 Molecular Weight (g/mol): 314.34 MDL Number: MFCD00059089 InChI Key: KYCAEIXHUXBNTQ-UHFFFAOYSA-N PubChem CID: 14086930 IUPAC Name: [2-(4-methoxyphenyl)acetyl] 2-(4-methoxyphenyl)acetate SMILES: COC1=CC=C(C=C1)CC(=O)OC(=O)CC2=CC=C(C=C2)OC