accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3952-66-7

IUPAC Name: methyl 2-oxobutanoate
Synonyms: dimethylpyruvic acid
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: XPIWVCAMONZQCP-UHFFFAOYSA-N
SMILES: CCC(=O)C(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)