accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3952-69-0

IUPAC Name: 5-hydroxy-4H-chromen-4-one
Synonyms: 5-hydroxychromen-4-one
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: CJMXMDVAKVSKFI-UHFFFAOYSA-N
SMILES: OC1=C2C(=O)C=COC2=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results