accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39520-24-6

IUPAC Name: 1,3-dimethyl 2-(2-methylpropyl)propanedioate
Synonyms: 1,3-dimethyl 2-(2-methylpropyl)propanedioate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: GAZFDPSEEIVCEX-UHFFFAOYSA-N
SMILES: COC(=O)C(CC(C)C)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results