accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)NH2ONH(S)NH2OOHO

CAS RN 39537-23-0

IUPAC Name: (2S)-2-[(2S)-2-aminopropanamido]-4-carbamoylbutanoic acid
Synonyms: alanyl glutamine
Molecular Weight (g/mol): 217.23
Molecular Formula: C8H15N3O4
InChi Key: HJCMDXDYPOUFDY-WHFBIAKZSA-N
SMILES: C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (6)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)