accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NONH

CAS RN 39546-32-2

IUPAC Name: piperidine-4-carboxamide
Synonyms: piperidine-4-carboxamide
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: DPBWFNDFMCCGGJ-UHFFFAOYSA-N
SMILES: NC(=O)C1CCNCC1
Molecular Weight (g/mol) 
  • (1)
  • (8)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)