accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2OHHCl

CAS RN 39547-15-4

IUPAC Name: 2-amino-5-methoxyphenol hydrochloride
Synonyms: 2-amino-5-methoxyphenol hydrochloride
Molecular Weight (g/mol): 175.61
Molecular Formula: C7H10ClNO2
InChi Key: ZXJIQOBYRBTOLH-UHFFFAOYSA-N
SMILES: Cl.COC1=CC(O)=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)