accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39557-45-4

IUPAC Name: 2-methylpropan-2-amine hydroiodide
Synonyms: erbumine hydroiodide
Molecular Weight (g/mol): 201.05
Molecular Formula: C4H12IN
InChi Key: NLJDBTZLVTWXRG-UHFFFAOYSA-N
SMILES: I.CC(C)(C)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results