accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3NCO

CAS RN 39570-63-3

IUPAC Name: methyl 2-isocyanato-4-methylpentanoate
Synonyms: methyl 2-isocyanato-4-methylpentanoate
Molecular Weight (g/mol): 171.2
Molecular Formula: C8H13NO3
InChi Key: UWFLIWPVRTVDNO-UHFFFAOYNA-N
SMILES: COC(=O)C(CC(C)C)N=C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results