CAS RN 39621-00-6
IUPAC Name:
2,6-dichloro-4-methylpyridine
Synonyms:
2,6-dichloro-4-methylpyridine
Molecular Weight (g/mol):
162.01
Molecular Formula:
C6H5Cl2N
InChi Key:
GTHRJKYVJZJPCF-UHFFFAOYSA-N
SMILES:
CC1=CC(Cl)=NC(Cl)=C1
1
–
1
of
1
results
1
Promotions Available
2,6-Dichloro-4-methylpyridine, 95%, Thermo Scientific Chemicals
2,6-Dichloro-4-methylpyridine, 95%, Thermo Scientific Chemicals
CAS: 39621-00-6 Molecular Formula: C6H5Cl2N Molecular Weight (g/mol): 162.013 MDL Number: MFCD09264302 InChI Key: GTHRJKYVJZJPCF-UHFFFAOYSA-N PubChem CID: 10898997 IUPAC Name: 2,6-dichloro-4-methylpyridine SMILES: CC1=CC(=NC(=C1)Cl)Cl