accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3964-54-3

IUPAC Name: N-(3-chloro-4-hydroxyphenyl)acetamide
Synonyms: N-(3-chloro-4-hydroxyphenyl)acetamide
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: JULKJDRBSRRBHT-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(O)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results