accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHOOHOOHHO

CAS RN 3969-84-4

IUPAC Name: 1-[5-(1,2-dihydroxyethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
Synonyms: 1-[5-(1,2-dihydroxyethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
Molecular Weight (g/mol): 222.24
Molecular Formula: C9H18O6
InChi Key: YCOMFYACDCWMMD-UHFFFAOYNA-N
SMILES: CC1(C)OC(C(O)CO)C(O1)C(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results