accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39711-79-0

IUPAC Name: N-ethyl-5-methyl-2-(propan-2-yl)cyclohexane-1-carboxamide
Synonyms: ethyl menthane carboxamide
Molecular Weight (g/mol): 211.35
Molecular Formula: C13H25NO
InChi Key: VUNOFAIHSALQQH-UHFFFAOYNA-N
SMILES: CCNC(=O)C1CC(C)CCC1C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results