accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCl

CAS RN 39718-00-8

IUPAC Name: 1-chloro-2-(methylsulfanyl)benzene
Synonyms: 1-chloro-2-(methylsulfanyl)benzene
Molecular Weight (g/mol): 158.64
Molecular Formula: C7H7ClS
InChi Key: IHLDFHCSSCVPQW-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)